Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syt9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003513487.1 | XP_003513535.1 | 3 - 4 | 991 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Probe spans exons |