Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rora | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007653916.1 | XP_007652106.1 | 7 - 8 | 994 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Nuclear Receptor 3' Most Probe spans exons |