Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007644909.1 | XP_007643099.1 | 2 - 3 | 3285 | 67 | |
XM_007644911.1 | XP_007643101.1 | 3 - 4 | 953 | 67 | ||
GenBank mRNA | JP061112.1 | - | 2 - 3 | 778 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Probe spans exons |