Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Npm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007648661.1 | XP_007646851.1 | 2 | 223 | 81 | |
GenBank mRNA | JI885143.1 | - | 2 | 223 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Both primers and probe map within a single exon |