Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cklf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003499679.2 | XP_003499727.1 | 1 - 2 | 234 | 95 | |
GenBank mRNA | JI863545.1 | - | 1 - 2 | 204 | 95 | |
JP042394.1 | - | 1 - 2 | 201 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Probe spans exons |