Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ticam2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003506772.2 | XP_003506820.1 | 1 | 219 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Both primers and probe map within a single exon |