Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gins2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007654858.1 | XP_007653048.1 | 1 - 2 | 402 | 80 | |
GenBank mRNA | JI883369.1 | - | 1 - 2 | 176 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Probe spans exons |