Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gins2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007654858.1 | XP_007653048.1 | 3 - 4 | 746 | 95 | |
GenBank mRNA | JI883369.1 | - | 3 - 4 | 520 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Probe spans exons |