Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003497836.2 | XP_003497884.2 | 2 - 3 | 320 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Protease Detect Genomic DNA Probe spans exons |