Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003497836.2 | XP_003497884.2 | 6 - 7 | 1000 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Protease 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |