Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fto | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003498014.2 | XP_003498062.1 | 1 - 2 | 106 | 79 | |
GenBank mRNA | JI880977.1 | - | 1 - 2 | 94 | 79 | |
JP048988.1 | - | 1 - 2 | 78 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Probe spans exons |