Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fto | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003498014.2 | XP_003498062.1 | 5 - 6 | 1030 | 98 | |
XM_007621141.1 | XP_007619331.1 | 5 - 6 | 1067 | 98 | ||
GenBank mRNA | JI880977.1 | - | 5 - 6 | 1018 | 98 | |
JP048988.1 | - | 5 - 6 | 1002 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Probe spans exons |