Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fto | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003498014.2 | XP_003498062.1 | 7 - 8 | 1296 | 103 | |
XM_007621141.1 | XP_007619331.1 | 7 - 8 | 1333 | 103 | ||
GenBank mRNA | JI880977.1 | - | 7 - 8 | 1284 | 103 | |
JP048988.1 | - | 7 - 8 | 1268 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |