Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Minpp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003507532.2 | XP_003507580.2 | 1 - 2 | 226 | 87 | |
GenBank mRNA | JP043847.1 | - | 1 - 2 | 226 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Probe spans exons |