Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Skor1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007641251.1 | XP_007639441.1 | 4 - 5 | 1321 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Probe spans exons |