Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptgfrn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007641376.1 | XP_007639566.1 | 2 - 3 | 444 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Probe spans exons |