Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abhd11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003515391.2 | XP_003515439.1 | 3 - 4 | 851 | 66 | |
GenBank mRNA | JI888602.1 | - | 3 - 4 | 478 | 66 | |
JP049246.1 | - | 3 - 4 | 478 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Probe spans exons |