Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem97 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003495808.2 | XP_003495856.2 | 1 - 2 | 194 | 53 | |
GenBank mRNA | JI870284.1 | - | 1 - 2 | 194 | 53 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Probe spans exons |