Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Carhsp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003496376.2 | XP_003496424.1 | 3 - 4 | 354 | 67 | |
XM_007649591.1 | XP_007647781.1 | 3 - 4 | 464 | 67 | ||
GenBank mRNA | JI893770.1 | - | 3 - 4 | 344 | 67 | |
JP060690.1 | - | 3 - 4 | 344 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |