Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chst15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003511990.1 | XP_003512038.1 | 2 - 3 | 889 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Probe spans exons |