Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007641137.1 | XP_007639327.1 | 1 - 2 | 364 | 62 | |
XM_007641138.1 | XP_007639328.1 | 1 - 2 | 365 | 62 | ||
XM_007641139.1 | XP_007639329.1 | 1 - 2 | 365 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Probe spans exons |