Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lysmd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003495052.2 | XP_003495100.2 | 2 - 3 | 402 | 92 | |
GenBank mRNA | JP046885.1 | - | 2 - 3 | 390 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Probe spans exons |