Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nme1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003501376.2 | XP_003501424.1 | 3 - 4 | 342 | 62 | |
XM_007642024.1 | XP_007640214.1 | 3 - 4 | 260 | 62 | ||
GenBank mRNA | JI885523.1 | - | 3 - 4 | 298 | 62 | |
JP038532.1 | - | 4 - 5 | 439 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Probe spans exons |