Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Casp14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003504698.1 | XP_003504746.1 | 5 - 6 | 628 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Protease 3' Most Detect Genomic DNA Probe spans exons |