Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chrna10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003509704.2 | XP_003509752.1 | 1 - 2 | 129 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Ion Channel Detect Genomic DNA Probe spans exons |