Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Batf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007651317.1 | XP_007649507.1 | 3 - 4 | 257 | 65 | |
XM_007651318.1 | XP_007649508.1 | 2 - 3 | 292 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Probe spans exons |