Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp1b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003497238.1 | XP_003497286.1 | 2 - 3 | 254 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Transporter Probe spans exons |