Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syt13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003497279.2 | XP_003497327.1 | 4 - 5 | 850 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Probe spans exons |