Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nudt16l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003506343.2 | XP_003506391.2 | 2 - 3 | 905 | 79 | |
GenBank mRNA | JI873449.1 | - | 2 - 3 | 444 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Probe spans exons |