Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Iffo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007652061.1 | XP_007650251.1 | 3 - 4 | 273 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Probe spans exons |