Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srpk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007649252.1 | XP_007647442.1 | 12 - 13 | 1680 | 60 | |
GenBank mRNA | JI896640.1 | - | 12 - 13 | 1680 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Protein Kinase Detect Genomic DNA Recommended Assay Probe spans exons |