Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rasa4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007641687.1 | XP_007639877.1 | 10 - 11 | 1227 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Probe spans exons |