Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007655136.1 | XP_007653326.1 | 2 - 3 | 463 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Probe spans exons |