Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H6pd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007651585.1 | XP_007649775.1 | 3 - 4 | 909 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Probe spans exons |