Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003498868.2 | XP_003498916.1 | 4 - 5 | 354 | 78 | |
XM_007634537.1 | XP_007632727.1 | 4 - 5 | 363 | 78 | ||
XM_007634543.1 | XP_007632733.1 | 4 - 5 | 442 | 78 | ||
XM_007634553.1 | XP_007632743.1 | 4 - 5 | 431 | 78 | ||
GenBank mRNA | JI873531.1 | - | 4 - 5 | 363 | 78 | |
JP050649.1 | - | 4 - 5 | 362 | 78 |