Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dusp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003504321.2 | XP_003504369.1 | 2 - 3 | 2544 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Protein Phosphatase 3' Most Detect Genomic DNA Probe spans exons |