Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003510167.2 | XP_003510215.1 | 2 - 3 | 183 | 77 | |
GenBank mRNA | JI891416.1 | - | 3 - 4 | 360 | 77 | |
JP045424.1 | - | 3 - 4 | 360 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |