Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003498216.2 | XP_003498264.1 | 3 - 4 | 648 | 65 | |
GenBank mRNA | JI876738.1 | - | 3 - 4 | 621 | 65 | |
JP052925.1 | - | 3 - 4 | 621 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Probe spans exons |