Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc45a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007653240.1 | XP_007651430.1 | 3 - 4 | 1353 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Probe spans exons |