Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Awat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003509812.1 | XP_003509860.1 | 3 - 4 | 262 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Probe spans exons |