Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab11b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003510502.2 | XP_003510550.1 | 4 - 5 | 605 | 99 | |
GenBank mRNA | JI895582.1 | - | 4 - 5 | 569 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Probe spans exons |