Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csrnp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003514117.2 | XP_003514165.1 | 3 - 4 | 1115 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Apoptosis Probe spans exons |