Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cryab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007629274.1 | XP_007627464.1 | 3 - 4 | 1269 | 66 | |
XM_007629280.1 | XP_007627470.1 | 2 - 3 | 1116 | 66 | ||
XM_007629284.1 | XP_007627474.1 | 3 - 4 | 1208 | 66 | ||
XM_007629291.1 | XP_007627481.1 | 3 - 4 | 790 | 66 | ||
XM_007629298.1 | XP_007627488.1 | 1 - 2 | 410 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Probe spans exons |