Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007646059.1 | XP_007644249.1 | 6 - 7 | 1334 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Probe spans exons |