Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nkx2-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007650520.1 | XP_007648710.1 | 6 - 7 | 1073 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Probe spans exons |