Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif4ebp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003498215.2 | XP_003498263.1 | 2 - 3 | 390 | 72 | |
GenBank mRNA | JI895765.1 | - | 2 - 3 | 349 | 72 | |
JP054409.1 | - | 2 - 3 | 350 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Probe spans exons |