Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003512675.2 | XP_003512723.1 | 2 - 3 | 4053 | 115 | |
GenBank mRNA | JI894597.1 | - | 2 - 3 | 3073 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |