Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtpbp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007654472.1 | XP_007652662.1 | 5 - 6 | 653 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Probe spans exons |