Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exog | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003505922.2 | XP_003505970.1 | 4 - 5 | 529 | 101 | |
GenBank mRNA | JI896513.1 | - | 3 - 4 | 366 | 101 | |
JP037634.1 | - | 3 - 4 | 366 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Apoptosis Probe spans exons |