Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ntan1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003502744.2 | XP_003502792.1 | 4 - 5 | 401 | 80 | |
XM_007643370.1 | XP_007641560.1 | 3 - 4 | 326 | 80 | ||
XM_007643371.1 | XP_007641561.1 | 4 - 5 | 404 | 80 | ||
GenBank mRNA | JI879919.1 | - | 2 - 3 | 179 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Probe spans exons |